Would you like to log in again?
Borosilicate Glass Culture Tubes, Globe Scientific
Supplier: Globe Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1510 1503 1522 1512 1517 1515 1505 1519
490015-330CS 147.45 USD InStock
490015-330 490015-332 490015-322 76264-512 490015-324 76264-510 490015-326 490015-328
Borosilicate Glass Culture Tubes, Globe Scientific
Tubes Culture Tubes
These tubes are made from superior quality borosilicate glass to reduce pH changes and contaminants that are potentially leached from soda lime glass tubes and are used for tissue culture and blood banks.
  • Excellent chemical resistance and durability
  • Features well-rounded bottoms and smooth fire-polished rims
TubeSaver™ packaging system helps prevent breakage and contamination during shipping and storage. Dimensional specifications are rigidly controlled for reliable, uniform results.
Certifications: An ISO 9001:2008 certified factory product.
Packaging: The tubes are neatly stacked in heavy-duty, shrink wrapped boxes.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty