Would you like to log in again?
LR3IGF-I Rapid ELISA Kit: Human, Biosensis
Supplier: Biosensis
BEK-2233-1P BEK-2233-2P
76264-592EA 826 USD InStock
76264-592 76264-590
LR3IGF-I Rapid ELISA Kit: Human, Biosensis
Assays ELISAs
The ELISA kit consists of a complete set of reagents and pre-coated plate to allow immediate assay of LR³IGF-I in culture media. Included in the kit are mouse monoclonal anti- LR³IGF-I capture antibody pre-coated onto an ELISA plate, standard LR³IGF-I protein, a biotinylated anti- LR³IGF-I detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin.
The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of LR³IGF-I present in samples and the supplied protein standard. This LR³IGF-I RapidTM ELISA kit has been developed, optimized and validated to quantify LR³IGF-I protein in cell culture medium.
It is likely to be used to measure LR³IGF-I in media and during downstream processing of media following a production cycle and is not intended for other use.
Delivery information: The ELISA kit box contains 1 or 2 x 96-well pre-coated strip plate, protein standards, detection reagents, wash and sample buffers, substrate buffer and detailed protocols.
Caution: This kit has been configured for research use only and is not to be used in diagnostic or clinical procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty