Would you like to log in again?
Phenylalanine Assay Kit (Colorimetric), BioVision
Supplier: BioVision
AB241000-100TEST
76265-744EA 789.07 USD InStock
76265-744
Phenylalanine Assay Kit (Colorimetric), BioVision
Assays Protein Assays
Colorimetric assay for quantifying phenylalanine (PHE) in biological samples.
  • Simple procedure takes less than 60 minutes
  • Fast and convenient
L-Phenylalanine (Phe) is an electrically-neutral amino acid, one of the twenty common and one of the three aromatic amino acids building the polypeptide backbone. Phenylalanine uses the same active transport channel as tryptophan to cross the blood-brain barrier, and, in large quantities, interferes with the production of serotonin. Errors in Phe metabolism lead to phenylketonuria or PKU which can have dire consequences.
Phenylalanine Assay Kit (Colorimetric) provides a quick, simple, accurate method for quantifying Phe in biological samples. In the assay, Phe is metabolized with the simultaneous formation of NADH which reacts with a probe to generate absorbance that can be followed colorimetrically at 450 nm. The assay is linear in the range from 0.2 to 10 nmol, allowing accurate determination of concentrations in the range of 1 - 50 µM phenylalanine.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty