Would you like to log in again?
EZClick™ Fucose (FucAz) Modified Glycoprotein Assay Kit (FACS/Microscopy), Green Fluorescence, BioVision Inc.
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB241020-100TEST
76265-788EA 857.74 USD InStock
76265-788
EZClick™ Fucose (FucAz) Modified Glycoprotein Assay Kit (FACS/Microscopy), Green Fluorescence, BioVision Inc.
Assays Protein Assays
Click-Chemistry based assay for measuring glycoproteins in biological samples such as intact cells.
  • Simple, fast, does not require lengthy incubation times
  • Includes sufficient materials for 100 assays in a 96-well plate format
Glycoproteins are grouped by the type of carbohydrate and amino acid linkage site. Fucose is found in glycoproteins and glycolipids present in vertebrates, invertebrates, plants, and bacteria. It is often a terminal sugar in glycans participating in cell–cell signaling, and migration implicated in physiological and pathological processes such as fertilization, embryogenesis, lymphocyte trafficking, immune responses, and cancer metastasis.
Most target proteins undergoing fucosylation are secretory or membrane proteins on the cell surface. Fucosylation is one of the most important modifications involved in cancer and inflammation. Several types of biomarkers containing fucose are linked to cancer e.g. alpha-fetoprotein (AFP) is used in the diagnosis of hepatocellular carcinoma and surface glycosphingolipid Globo H is an epitope found on the cell surface of breast, prostate, and ovarian cancers. Since glycoproteins are not directly encoded in the genome, methods of their characterization and analyses are of great interest.
EZClick™ Fucose (FucAz) Modified Glycoprotein Assay Kit, a highly specific, simple and robust method for labeling and detection of fucosylated proteins within cells. We use fucose analog that is fed directly into the cells, processed via fucose salvage pathway and incorporated into the glycoproteins. Followed by click reaction with alkyne-containing dye, this system offers a powerful method for imaging the localization, trafficking, and dynamics of glycans, or detection by FACS for quantitative studies. Fucose-labeled glycoproteins can be directly detected in 1D or 2D gels using the appropriate excitation sources, or enriched by immunoprecipitation with biotin-alkyne or antibodies prior to proteomic analysis.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty