Would you like to log in again?
ITS Animal Free™ - (Insulin-Transferrin-Selenium) Media Supplement, InVitria
Supplier: InVitria
777ITS091-10ML 777ITS091100ML 777ITS09110X10
76266-356EA 1128.85 USD InStock
76266-356 76266-354 76266-352
ITS Animal Free™ - (Insulin-Transferrin-Selenium) Media Supplement, InVitria
Cell Culture Supplements and Additives
InVitria’s ITS Animal-Free™ is formulated to reduce serum from cell culture media.
InVitria’s ITS Animal-Free™ (Insulin-Transferrin-Selenium) is a recombinant, blood-free cell culture media supplement used to reduce serum and serum-derived components from cell culture media while optimizing cell growth. InVitria offers a family of defined supplements that supply the optimal amounts of the essential protein components in defined, recombinant form: ITS, ITSE, ITSE+A Animal Free. Our recombinant insulin-transferrin supplements are recommended for use with most cultured cell types, including: Hybridoma, CHO, VERO, HEK293, stem cells, fibroblasts, primary cell isolation and passage, and immortalized cell lines.
Made in the USA.
Certifications: cGMP grade.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty