Would you like to log in again?
PALL Filter Element Replacement, Filtermart
Supplier: FILTERMART TE
POC1001SU
76267-644EA 192.26 USD InStock
76267-644
PALL Filter Element Replacement, Filtermart
Filters Gas Filters
Particulate filters, also known as after-filters, are used to remove solid particulate contamination from compressed air line.
They are typically located after a compressed air desiccant dryer. These compressed air particulate filters provide the highest level of clean compressed air with a minimum loss of pressure. Compressed air particulate filter elements are tough enough to hold their shape under pressure and maintain even pressure differentials to avoid collapsing of the filter element.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty