Would you like to log in again?
Flex500™ High-Temperature Masking Kit, Caplugs
Supplier: Caplugs
MKITHVC-062-281
76267-698EA 2563.4 USD InStock
76267-698
Flex500™ High-Temperature Masking Kit, Caplugs
Fittings and Connectors
MKITS HVC Series High-Temperature Masking Kits allow you to keep a wide variety of sizes and styles on hand for quick and easy access.
  • Flex500™ material can withstand temperatures up to 500 °F
MKIT caps are ideal for masking applications including powder coating, e-coating, painting and more.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty