Would you like to log in again?
Flame Resistant Hoods, Balaclava, Zoro
Supplier: Zoro
H11RY H31NK
76253-886EA 100.43 USD InStock
76253-886 76267-818
Flame Resistant Hoods, Balaclava, Zoro
Workwear
Over the head flame resistant hood covers head, face and neck.
  • ATPV rating 12.0 cal./cm2
Certifications: Complies with Hazard Risk Category (HRC) 2, Standards ASTM F1506, OSHA 1910.269, NFPA 70E, CSA Z462, UL Certified to NFPA 2112.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty