Would you like to log in again?
Anti-Static Cleanroom Marking Tapes, UltraTape Industries
Supplier: UltraTape
2265WH300P3D 2265WH200-P3D 2265WH100-P3D
76268-024EA 118.48 USD InStock
76268-024 76248-426 76248-210
Anti-Static Cleanroom Marking Tapes, UltraTape Industries
Tapes
Cleanroom Anti-static tape available in for floor, tool and safety marking as well as a wide range of applications were opaque colors are needed. Printed and overlaminated for extra durability and solvent resistance. Bright opaque colors and black printed text.
  • Excellent durability
  • Anti-static and cleanroom compatible
  • Solvent resistant
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty