Would you like to log in again?
MPC Series Couplings, Medical Grade, Colder Products Company
Supplier: CPC (Colder Products Company)
MPC17006T39B25 MPC17002T03 MPC17004T03B25 MPC17006T03
76247-744PK 138.04 USD InStock
76247-744 76253-258 76247-708 76268-306
MPC Series Couplings, Medical Grade, Colder Products Company
Fittings and Connectors
MPC Series couplings add ease of use and security to your most critical fluid handling applications.
  • Easy to operate
  • BSE/TSE free materials
  • Ergonomic thumb latch
  • Sterilizable by autoclave, Et0, e-beam or gamma
These couplings provide optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.
Operating temperature: −40 to 121 °C (for polycarbonate) and −40 to 149 °C (for polysulfone)
Certifications: USP Class VI, ADCF certified and meets biocompatibility requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty