Would you like to log in again?
Neoprene Gloves, MAPA Professional
Supplier: Mapa Professionnel
N-740
76268-332CS 587.01 USD InStock
76268-332
Neoprene Gloves, MAPA Professional
Gloves
Ultra-high performance chemical protection.
  • Good grip, with embossed texture
  • Easy to pull on and off with chlorinated interior
Broad chemical resistance (acids, aliphatic solvents, solvent mixtures), increased by the thickness of the material.
The two neoprene layers superimposed increase the mechanical and chemical resistance. The glove red inner layer ensures that any wear is clearly visible, providing optimal safety for the user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty