Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SYBR® Green I ≥98% (by HPLC) fluorescent dye

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Thiazole Green

AB285424-5MG AB285424-25MG
76268-858EA 2183.33 USD InStock
76268-858 76268-856
SYBR® Green I ≥98% (by HPLC) fluorescent dye
SYBR® Green I
SYBR Green I is a highly sensitive fluorescent DNA binding stain for staining of double stranded DNA (dsDNA) or oligonucleotides in agarose or polyacrylamide gels.

SYBR is a registered trademark of Thermo Fisher Scientific.


Formula: C₃₂H₃₇N₄S
MW: 509.73 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00284715
CAS Number: 163795-75-3
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty