Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Trimming Blades, Avantik
Supplier: Avantik
GR2094-B GR2094-A
76270-460CS 369.53 USD InStock
76270-460 76270-144
Trimming Blades, Avantik
Scalpels and Scalpel Assemblies Scalpel Blades
Stainless steel trim blades are perfect for use in the pathology lab for hard tissues with fibers, soft, or semi-fixed organs.
  • Two sizes available
  • Convenient storage box keeps blades safe and sharp
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty