Would you like to log in again?
Aqueous Clarifier - Optik Type 2, Avantik
Supplier: Avantik
RS4366-B
76270-180CS 971.32 USD InStock
76270-180
Aqueous Clarifier - Optik Type 2, Avantik
Stains Microscopy and staining reagents
Optik Type 2 Aqueous Clarifier is designed to be used with Type 2 Hematoxylin to produce cell transparency and remove any non-specific staining. Slightly stronger clarifier than the Optik Alcoholic Clarifier.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty