Would you like to log in again?
Stackable Metal Slide Cabinets, Avantik
Supplier: Avantik
ST7077 ST7076
76270-352EA 261.19 USD InStock
76270-352 76270-350
Stackable Metal Slide Cabinets, Avantik
Microscope Slide Boxes
Holds up to 5000 standard 3×1" microscope slides.
  • Interlocking, stackable to 10 cabinets high
  • Cabinet and base are stackable with the Boekel Cabinet
  • Cabinet base features rubber feet to prevent scratching and slipping
Cabinet is supplied with a two-year warranty.
Delivery information: Includes plastic drawer dividers, sponge blocks spaces and identification labels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty