Would you like to log in again?
SureSlide™ Standard Microscope Slides, Avantik
Supplier: Avantik
SL6333-3 SL6333-9 SL6333-6 SL6333-2 SL6333-1 SL6333-5 SL6333-4 SL6333-7
76270-484CS 504.91 USD InStock
76270-484 76270-472 76270-482 76270-480 76305-454 76270-478 76270-476 76270-474
SureSlide™ Standard Microscope Slides, Avantik
Slides Microscope Slides
SureSlide™ microscope slides provide consistent and reliable results.
  • Optimized to ensure consistent barcode scanning
  • 100% quality guarantee
These slides are compatible with all major manufacturer automated printers; resulting in superior print quality.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty