Would you like to log in again?
SureBond™ Charged Clipped Corner Microscope Slides, Avantik
Supplier: Avantik
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SL6335-4 SL6335-9 SL6335-3 SL6335-2 SL6335-1 SL6335-5
76270-494CS 1085.13 USD InStock
76270-494 76270-492 76270-502 76270-500 76270-498 76270-496
SureBond™ Charged Clipped Corner Microscope Slides, Avantik
Slides Adhesive Slides
SureBond™ microscope slides provide consistent and reliable results.
  • Optimized to ensure consistent barcode scanning
  • High-quality charge ensures excellent adhesion to eliminate tissue loss
  • 100% quality guarantee
These slides are compatible with all major manufacturer automated printers; resulting in superior print quality.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty