Would you like to log in again?
SureBond™ 2 IP Paint Microscope Slides, Avantik
Supplier: Avantik
SL6349-7 SL6349-1 SL6349-3 SL6349-4 SL6349-2 SL6349-5
76270-518CS 1431.64 USD InStock
76270-518 76270-516 76270-514 76270-524 76270-522 76270-520
SureBond™ 2 IP Paint Microscope Slides, Avantik
Slides Microscope Slides
SureBond™ microscope slides provide consistent and reliable results.
  • Optimized to ensure consistent barcode scanning
  • High-quality charge ensures excellent adhesion to eliminate tissue loss
  • 100% quality guarantee
These slides are compatible with all major manufacturer automated printers; resulting in superior print quality.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty