Would you like to log in again?
Twin Bucket, Contec®
Supplier: Contec
2743S
76270-914CS 330.59 USD InStock
76270-914
Twin Bucket, Contec®
Buckets
The Twin Bucket is a divided bucket with a curved stainless steel wringer.
  • Ideal for smaller controlled environments and labs with limited storage space
  • Designed to work with the curved stainless steel wringer
  • Fully autoclavable (casters are non-autoclavable)
The Twin Bucket is made of durable polypropylene, with a well -fitting lid to eliminate liquid spillage during transport. Use one side of the bucket for cleaning and disinfecting solutions, and the other for rinsing.
Delivery information: Includes 4 casters for added mobility.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty