Would you like to log in again?
Epigenetics Screening Library, Cayman Chemical Company
Supplier: Cayman Chemical
11076-50 11076-100
76270-988EA 2278.25 USD InStock
76270-988 76270-986
Epigenetics Screening Library, Cayman Chemical Company
Assays Cellular Assays
For screening a variety of epigenetic compounds
  • Contains >140 small molecules
  • Includes compounds that modulate the activity of methyltransferases, demethylases, HATs, HDACs, and acetylated lysine reader proteins
The Epigenetics Screening Library contains more than 140 small molecules that are known to modulate the activity of a variety of epigenetic ‘writers and erasers’ and ‘reader’ proteins in a 96-well Matrix tube rack format as 10 mM stocks in DMSO. It may include compounds that modulate the activity of methyltransferases, demethylases, histone acetyltransferases, histone deacetylases, and acetylated histone binding proteins. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty