Would you like to log in again?
Prostaglandin Screening Library II, Cayman Chemical
Supplier: Cayman Chemical
10502-50
76271-060EA 1399.07 USD InStock
76271-060
Prostaglandin Screening Library II, Cayman Chemical
Assays Cellular Assays
For D and E series prostaglandin analog screening
  • Contains >60 compounds
  • Includes the “D- and E-series” prostaglandins
The Prostaglandin Screening Library II contains more than 60 “D and E-series” of prostaglandins in a 96-well Matrix tube rack format as 2 mM stocks in DMSO. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty