Would you like to log in again?
Fatty Acid Screening Library, Cayman Chemical Company
Supplier: Cayman Chemical
10504-50 10504-100
76271-084EA 1765.27 USD InStock
76271-084 76271-086
Fatty Acid Screening Library, Cayman Chemical Company
Assays Cellular Assays
For screening of variety of fatty acids
  • Contains >75 compounds
  • Includes arachidonic, linoleic, eicosapentaenoic, stearic, α- and γ-linolenic, oleic, and palmitic acids
The Fatty Acid Screening Library contains more than 75 compounds of fatty acids with diverse biological activities in a 96-<wbr/>well Matrix tube rack format as 2 mM stocks in DMSO. The library may also include some of the most commonly-ordered fatty acids, including arachidonic, linoleic, eicosapentaenoic, stearic, α- and γ-linolenic, oleic, and palmitic acids. As well as some unusual fatty acids, including 9-thiastearic acid, 2-hydroxymyristic acid, AUDA, and CUDA may be included. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty