Would you like to log in again?
Corepoint Scientific™ White Diamond Series Undercounter and Countertop Refrigerators and Freezers, Horizon Scientific
Supplier: Horizon Scientific
PR011WWWLH/0 PF051WWW/0A PR061WWW/0 PR031WWW/0 FF051WWW/0M PR051WWWADALH0 PF051SSSLH/0M PF051SSS/0M PR051SSS/0 PR051WWGADALH0 PF051WWW/0M PR051WWW/0 PR011WWW/0 PF051WWWLH/0A PR051WWG/0 PF021WWWLH/0A PR051WWGADA/0 PR051WWGLH/0 PF051WWW/0M30 PR031WWG/0 PR051WWWLH/0 PR051WWWADA/0 PF021WWWLH/0M PF051WWWADAL0M PF021WWW/0M PR051SSSLH/0 PR011WWG/0 PF051WWWLH/0M PR011WWGLH/0 FR051WWW/0 PR061WWG/0 PF051WWW/0M40 PF051WWWADA/0M PF021WWW/0A
76271-294EA 2030.34 USD InStock
76271-294 76271-296 76271-290 76271-292 76271-316 76271-338 76271-318 76271-356 76271-312 76271-334 76271-314 76271-336 76271-298 76271-352 76271-330 76271-310 76271-354 76271-332 76271-340 76271-328 76271-306 76271-308 76271-302 76271-324 76271-346 76271-304 76271-326 76271-320 76271-342 76271-286 76271-322 76271-344 76271-300 76271-288
Corepoint Scientific™ White Diamond Series Undercounter and Countertop Refrigerators and Freezers, Horizon Scientific
Refrigerators
These undercounter and countertop refrigerators and freezers provide premium laboratory cold storage for scientific and medical use
  • Integrated temperature alarms
  • Precise microprocessor temperature control
  • Forced draft circulation
  • Keyed door locks
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
Freestanding units require a 4-inch margin on all sides for adequate ventilation. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Select models offer auto defrost cycles, ADA compliance, and left hinged options. For lower temperature units, -30 and -40 °C models are also available.
Certifications: *Select models ENERGY STAR certified: 76271-318, 76271-320, 76271-324, 76271-326, 76271-322, 76271-328, 76271-352, 76271-330, 76271-286, 76271-288, 76271-294, 76271-296, 76271-298, 76271-300, 76271-354, 76271-356, 76271-290, 76271-292, 76271-332, 76271-334, 76271-336, 76271-338, 76271-302, 76271-304, 76271-306, 76271-308, 76271-310, 76271-312, 76271-340, and 76271-342
Ordering information: All units have a three-year parts and labor warranty. Select models come with an additional two-year compressor parts warranty. (Please refer to unit datasheets to confirm).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty