Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CALR Mouse Monoclonal Antibody (PE) [clone: FMC 75]

Supplier: Enzo Life Sciences
ABS668PE-0200
76271-382EA 840 USD InStock
76271-382
Anti-CALR Mouse Monoclonal Antibody (PE) [clone: FMC 75]
Antibodies
Recommended Applications: Flow Cytometry

Type: Primary
Antigen: CALR
Clonality: Monoclonal
Clone: FMC 75
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1κ
Reactivity: Human;Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty