Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CKAP4 Mouse Monoclonal Antibody [clone: G1/296]

Supplier: Enzo Life Sciences
ABS669-0100
76271-384EA 519 USD InStock
76271-384
Anti-CKAP4 Mouse Monoclonal Antibody [clone: G1/296]
Antibodies
Recommended Applications: ELISA, Flow Cytometry, ICC, IHC (FS), IP, WB

Type: Primary
Antigen: CKAP4
Clonality: Monoclonal
Clone: G1/296
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2aκ
Reactivity: Human;Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty