Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP90 Mouse Monoclonal Antibody [clone: AC88]

Supplier: Enzo Life Sciences
ABS670-0200
76271-386EA 705 USD InStock
76271-386
Anti-HSP90 Mouse Monoclonal Antibody [clone: AC88]
Antibodies
Recommended Applications: IHC (PS), WB

Type: Primary
Antigen: HSP90
Clonality: Monoclonal
Clone: AC88
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1κ
Reactivity: Human;Mouse;Rat;Fusobacterium nucleatum;Water mold;Sheep;Scallop;Rabbit;Porcine;Mussel;Monkey;Mycobacterium leprae;Hamster;Guinea pig;Fish;E. coli;Dog;Chicken;C. elegans;Beluga

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty