Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT Rabbit Monoclonal Antibody [clone: RM316]

Supplier: Enzo Life Sciences
ABS674-0100
76271-436EA 622 USD InStock
76271-436
Anti-AKT Rabbit Monoclonal Antibody [clone: RM316]
Antibodies
Recombinant monoclonal antibody (Rabbit IgG).

Recommended Applications: IHC (PS), WB

Type: Primary
Antigen: AKT
Clonality: Monoclonal
Clone: RM316
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty