Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KDEL Mouse Monoclonal Antibody (DyLight™ 488) [clone: 10C3]

Supplier: Enzo Life Sciences
ABS679488-0050
76271-446EA 669 USD InStock
76271-446
Anti-KDEL Mouse Monoclonal Antibody (DyLight™ 488) [clone: 10C3]
Antibodies
Recommended Applications: Flow Cytometry

Type: Primary
Antigen: KDEL
Clonality: Monoclonal
Clone: 10C3
Conjugation: DyLight™ 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Yeast;Plant;Mammals;Insect;Avian

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty