Would you like to log in again?
LABEL PIGGYBACK 4X2IN WHITE 3IN CORE RL1
Supplier: GA INTERNATIONAL INC
Label Piggyback 4X2In White 3In Core RL1^ Label, Autoclave Piggyback, Dimension: 4IN x 2IN (2 Label of 2.5in x 1.75in & 4, Dimension: of 1.25IN x 0.25in ) White, 3in Core, 500 Label Sets/Roll, With Perforation, Removable
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LABEL PIGGYBACK 4X2IN WHITE 3IN CORE RL1 76273-690 807656015866 Roll of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty