Would you like to log in again?
CRYOLABEL RED 1.125X1+1.595IN WRAP RL1
Supplier: GA INTERNATIONAL INC
Cryolabel Red 1.125X1+1.595In Wrap RL1^ CryoLabel, Deep-freeze, Thermal transfer, wrap around, Dimension: 1.125x1+1.595in wrap, Color: red + transparent, Core: 1in, Mark: notch, without perforation, For vials and tubes in long-term cryo storage and printe
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRYOLABEL RED 1.125X1+1.595IN WRAP RL1 76273-998 807656017532 Roll of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty