Would you like to log in again?
CRYOLABEL 1IN WH 1.34X1+1.38IN WRAP RL1
Supplier: GA INTERNATIONAL INC
Cryolabel 1In Wh 1.34X1+1.38In Wrap RL1^ CryoLabel, Deep-freeze, Thermal transfer, wrap around, Dimension: 1.34x1+1.38in wrap, Color: white + transparent, Core: 1in, Mark: notch, Perforated, For vials and tubes in long-term cryo storage and printers, Labe
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRYOLABEL 1IN WH 1.34X1+1.38IN WRAP RL1 76274-044 807656017747 Roll of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty