Would you like to log in again?
CRYOLABEL 1X0.625+1IN WRAP 3IN CORE RL1
Supplier: GA INTERNATIONAL INC
Cryolabel 1X0.625+1In Wrap 3In Core RL1^ CryoLabel, Cryo & Autoclave Wrap Around, Dimension: 1in x 0.625in + 1in wrap / 25.4mm x 15.9mm + 25.4mm wrap, 3in Core, Label/Roll: 4000, Perforated, Mark: Notch
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRYOLABEL 1X0.625+1IN WRAP 3IN CORE RL1 76274-058 807656027906 Roll of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty