Would you like to log in again?
CRYOLABEL 1X1.18IN+1.69IN WRAP 3IN RL1
Supplier: GA INTERNATIONAL INC
Cryolabel 1X1.18In+1.69In Wrap 3In RL1^ CryoLabel, Cryo & Autoclave Wrap Around, Dimension: 1in x 1.18in + 1.69in wrap / 25.4mm x 29.97mm + 42.93mm wrap Core: 3in (76.2mm), Label/Roll: 3,000 Color: White+Transparent Mark: Notch
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRYOLABEL 1X1.18IN+1.69IN WRAP 3IN RL1 76274-062 807656017808 Roll of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty