Would you like to log in again?
CRYOLABEL 1IN TRANSPARENT 4INX2IN ROLL 1
Supplier: GA INTERNATIONAL INC
Cryolabel 1in Transparent 4inx2in Roll 1^ Deep-Freeze Omni-Tag* CryoLabel, Core: 1in, Color: transparent, with perforation, Mark: Notch, Labels/Roll: 500, Dimension: 4in x 2in, for barcode and thermal printers. Withstand long-term storage in Deep-freeze a
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRYOLABEL 1IN TRANSPARENT 4INX2IN ROLL 1 76274-218 807656018539 Roll of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty