Would you like to log in again?
BLACKOUT LABEL WHITE 1IN 0.9375IN 1 ROLL
Supplier: GA INTERNATIONAL INC
Blackout Label White 1In 0.9375In 1 Roll^ Blackout Label, Color: White, Perforation: Yes, Adhesive: Removable, Designed to fit microscope slides, Durable, Made of Waterproof thermoplastic film, and leave no residue behind, Chemical resistant, Core diamete
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BLACKOUT LABEL WHITE 1IN 0.9375IN 1 ROLL 76274-488 807656031606 Roll of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty