Would you like to log in again?
CRYOGENIC TAPE WHITE 1X0.75INX50FT 1ROLL
Supplier: GA INTERNATIONAL INC
Cryogenic Tape White 1X0.75Inx50Ft 1Roll^ TAMPER-PROOF* Cryogenic Tape, Destructible, Color: White, For sealing containers and other items, Capable of withstanding cryogenic storage environments, Water-proof, Temperature range: from -196 to 120 deg C, Cor
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRYOGENIC TAPE WHITE 1X0.75INX50FT 1ROLL 76275-996 807656084152 Roll of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty