Would you like to log in again?
ProPac™ Elite WCX HPLC Columns
Supplier: Thermo Scientific
ProPac™
302977 303028 302976 303026 303027 302972 303355 302973
76276-284EA 1753.34 USD InStock
76276-284 76276-282 76276-288 76276-276 76477-166 76276-278 76276-280 76276-290
ProPac™ Elite WCX HPLC Columns
Chromatography Columns
Thermo Scientific™ ProPac™ Elite WCX weak cation exchange (WCX) liquid chromatography column.
  • Superior resolution power for proteins, monoclonal antibodies, and associated charge variants
  • High efficiency with reproducible separations
  • High recovery with low carry-over
  • Wide pH operating range: 2 to 12
  • High temperature stability: up to 60 °C
  • High throughput
  • Compatible with CX-1 pH gradient buffers
Designed for protein characterization including therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs) and biosimilar proteins, this column offers high resolution, high efficiency separation of proteins and their charge variants. The unique column chemistry provides excellent performance under a broad range of pH, temperature and mobile phase compositions with excellent recovery and low carryover.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty