Would you like to log in again?
COLUMN WCX PROPAC 3 COLUMN 1 LOT 3/ PACK
Supplier: Thermo Scientific
Propac* Column Wcx 3 Column 1 Lot 3/ PACK^ Propac* Elite WCX Column, Analytical, Pore Size: 5um, 3 Columns from 1 lot, Has a carboxylate functional group, Characterization and quality control assessment of monoclonal antibodies, High-resolution separations with sample recovery, Dimension: 4x150mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN WCX PROPAC 3 COLUMN 1 LOT 3/ PACK 76276-288 302976 Pack of 3 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty