Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dolly for 8500 Series Stack Chairs, 660 lbs. Weight Capacity, Black Powder Coated Steel, National Public Seating

Supplier: National Public Seating
DY85
76276-954EA 564.32 USD InStock
76276-954
Dolly for 8500 Series Stack Chairs, 660 lbs. Weight Capacity, Black Powder Coated Steel, National Public Seating
Carts
Industrial strength steel stack chair dolly for all your commercial storage and transport needs.

  • Dolly can be used with chairs 31H×19³/₄W×22" D
  • Constructed of 11-gauge steel
  • 4Ø×1¹/₄W" polyurethane casters
  • 165 lbs. per caster weight capacity
  • Chairs load at an angle to maximize stack ability
  • Black powder coated steel
  • Stacks up to 40 of the 8500 series

The DY-85 chair dolly, an 11-gauge steel unit, loads at an angle to securely and impressively hold up to 40 of the NPS 8500 Series stack chairs for easy transport and storage. Features black powder-coated paint finish that will not chip and four polyurethane casters (two that swivel) for easy mobility over nearly any surface. Each caster boasts a 165 lbs. weight capacity for a total of 660 lbs.

Can be used with chairs 76276-694, 76276-696, 76276-698, and 76276-700.

Certifications: MAS Certified Green.

Ordering information: Ten years warranty.

Delivery information: 48 hours QuickShip.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty