Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NPS® Folding Table Dollys for Horizontal Storage, National Public Seating

Supplier: National Public Seating
DY-3072 DY-3096
76276-968EA 735.52 USD InStock
76276-968 76276-970
NPS® Folding Table Dollys for Horizontal Storage, National Public Seating
Carts
High-quality folding table dolly is of a great value.

  • Holds 4, 5, 6, or 8' tables
  • Laid flat, stacks up to 12 tables high
  • Rolls on 4 casters: 2 swivel and 2 stationary
  • Dark brown powder-coated steel
  • Total weight capacity: 1100 lbs.
  • Casters are 4×11/4" and made of nylon

The DY-3072 Dolly stores and moves folding tables up to 6' long with total ease. Boasts a 1100 lbs. weight capacity and the brown scratch-resistant powder-coated paint finish resists chipping even after years of rigorous use. Two of the four nylon casters swivel for easy maneuvering. Each caster boasts a 275 lbs. weight capacity for a total of 1100 lbs.

Certifications: MAS certified Green.

Delivery information: Supplied with a ten year warranty.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty