Would you like to log in again?
Electrical Tape 56, Polyester Film, Tapecase
Supplier: TAPECASE LTD. TE
56-YELLOW-1/4
76277-182EA 61.96 USD InStock
76277-182
Electrical Tape 56, Polyester Film, Tapecase
Tapes
Polyester Film Electrical Tape 56 is a general purpose polyester tape with thermosetting rubber adhesive.
  • Exhibits excellent chemical, solvent and moisture resistance
  • Resists cut-through and abrasion
Designed for insulating applications requiring a thin, durable tape with high dielectric strength. It can withstand higher-temperature conditions than tapes with acetate cloth backings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty