Would you like to log in again?
Biohazard Waste Bags, Autoclavable, Medegen Medical Products
Supplier: Medegen Medical Products
5012 5119 872 8-150 844 842 881 5138 5055.HTS 5034 5001.4 5142.F 858 5020.F 5028 5053 884 882 854 5010 880 855 8-300 5013 856 8-702 852 8-250 848 2120 846 864
76380-308CS 104.72 USD InStock
76380-308 76380-306 76380-300 76380-268 76380-266 76380-288 76380-304 89237-572 76380-260 76380-282 76380-280 76380-264 76277-262 76380-286 76380-284 76380-262 76238-204 76238-242 76380-256 76380-278 76380-312 76380-276 76380-298 76380-254 76380-310 76380-258 76380-314 76380-270 76380-292 76380-296 76380-272 76380-294
Biohazard Waste Bags, Autoclavable, Medegen Medical Products
Bags Waste Disposal Bags
These autoclave bags are built to withstand the rigors of steam sterilization while meeting the requirements for the collection, storage, transport and processing of biohazardous waste.
  • Standard autoclave bags are tested to exceed 165 g of dart impact for compliance in the majority of US states
  • Biohazardous labeling in both English and Spanish
  • International Biohazard Symbol in easy-to-see contrasting color
Delivery information: Twist ties included.
Packaging: Flat pack.
Caution: Not for use in CA, FL or PA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty