Would you like to log in again?
Single Tool Control Power Supply for Electric Screwdrivers, ASG
Supplier: ASG, Division of Jergens
PS-55
76277-272EA 740.15 USD InStock
76277-272
Single Tool Control Power Supply for Electric Screwdrivers, ASG
Power Supplies
Switchable two-speed output delivers a full 30 VDC on 'HI' setting – extra voltage when needed for optimal performance.
  • Able to use with BL series with torque up to 15 lbf.in
  • Standard clutch signal output, can be used with counters or PLC’s
  • Circuit breaker protected
Suitable for use with A, CL, SS, TL, BLG-4000 and BLG-5000 series tools. Not compatible with A-6500, A-6500HT, CL-7000HT or CL-9000.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty