Would you like to log in again?
Tissue Block and Slide Storage Boxes, Newcomer Supply
Supplier: Newcomer Supply
6875 6877
76278-818CS 155 USD InStock
76278-818 76278-816
Tissue Block and Slide Storage Boxes, Newcomer Supply
Microscope Slide Boxes
Stackable, corrugated cardboard file boxes for slide or block storage.
  • Affordable and easy to use storage systems for slides and blocks
  • Boxes are stackable
  • Recording area on the front panel of each box
  • Corrugated cardboard that supports up to 250 lbs.
  • Dimensions 12W×33/4H×111/2L" (storage box)
The cardboard storage systems are an economical way to store both slides and blocks. These file boxes are sturdy enough for transport and storage and are stackable. There is a well organized and detailed recording area on front panel of each box to easily identify the slides or blocks contained in the box.
Made in the U.S.A.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty