Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNF Human Monoclonal Antibody [clone: CDP 571 (Humicade)]

Supplier: Enzo Life Sciences
ABS564-0200
76284-580EA 622 USD InStock
76284-580
Anti-TNF Human Monoclonal Antibody [clone: CDP 571 (Humicade)]
Antibodies
Recombinant monclonal antibody (Human IgG4κ).

Recommended Applications: ELISA

Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: CDP 571 (Humicade)
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG4κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty