Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Fab fragment kappa Isotype Control [clone: MOPC-21]

Supplier: Enzo Life Sciences
ABS570-0200
76284-592EA 633 USD InStock
76284-592
Human Fab fragment kappa Isotype Control [clone: MOPC-21]
Antibodies
Recombinant monclonal antibody (Human Fab fragment κ).

Recommended Applications: Flow Cytometry, WB

Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: Fab fragment kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty