Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD3E Rat Monoclonal Antibody [clone: YTH 12.5]

Supplier: Enzo Life Sciences
ABS623-0200
76284-608EA 633 USD InStock
76284-608
Anti-CD3E Rat Monoclonal Antibody [clone: YTH 12.5]
Antibodies
Recombinant monclonal antibody (Rat IgG2bλ).

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: YTH 12.5
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2bλ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty