Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD34 Mouse Monoclonal Antibody [clone: My10]

Supplier: Enzo Life Sciences
ABS627-0200
76284-616EA 622 USD InStock
76284-616
Anti-CD34 Mouse Monoclonal Antibody [clone: My10]
Antibodies
Recombinant monclonal antibody (Mouse IgG1κ).

Recommended Applications: Flow Cytometry, IHC, IP

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: My10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty