Would you like to log in again?
CRYO.S BIOBANKING TUBE RED 1000 UL CS960
Supplier: Greiner Bio-One
Cryo.S Biobanking Tube Red 1000 Ul CS960^ CRYO.S Biobanking Tube, 2D codes, Rack with 96 Red Screw capped tubes, Sterile, 1 Manual capping tool, Datamatrix barcode (generic 2D sequence), 2 racks/bag, Case of 960, Size: 1000ul
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRYO.S BIOBANKING TUBE RED 1000 UL CS960 76284-630 978563 Case of 960 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty