Would you like to log in again?
Elite Series Waterproof Conductivity Pocket Testers, Thermo Scientific
Supplier: Thermo Scientific
Elite
ELITECTSCUP ELITECTSPIN
76284-652EA 148.68 USD InStock
76284-652 76284-654
Elite Series Waterproof Conductivity Pocket Testers, Thermo Scientific
Electrochemical Meters Conductivity Meters
The Elite CTS pocket testers are ideal for flexible on-the-go conductivity, TDS, salinity and temperature testing in a compact, travel-ready design. Available with pin-style direct-measure sensor or with cup-style fillable sensor.
  • Flexible testing option
  • Measure conductivity, TDS or salinity with temperature
  • Large back-lit graphic display
  • Use 4 x AAA alkaline batteries
  • Average 150 hours of battery life
  • Wide leak-proof cap serves as stable base
  • Replaceable sensors
Ideal for testing beverages including homebrew beer and wine, food, hydroponics, aquaculture and aquariums, environmental monitoring and more.
Certifications: Meets IP67 waterproof rating.
Ordering information: Supplied with batteries, lanyard, soft carry case.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty